Merck
  • Antimicrobial activities of components of the glandular secretions of leaf cutting ants of the genus Atta.
Antimicrobial activities of components of the glandular secretions of leaf cutting ants of the genus Atta.
Antonie van Leeuwenhoek (2009-02-14)
Adriana de Lima Mendonça, Carlos Eduardo da Silva, Fernando Lucas Torres de Mesquita, Rousseau da Silva Campos, Ruth R Do Nascimento, Eulália Camelo Pessoa de Azevedo Ximenes, Antônio Euzébio G Sant'Ana
ABSTRACT
The secretions of the mandibular and metapleural glands of leaf cutting ants contain antimicrobial substances that protect the mutualistic fungal colony within the nest from attack by parasitic micro-organisms. The major constituents of these secretions (citral, 4-methyl-3-heptanol, 2-heptanone, 3-octanone, 4-methyl-2-heptanone, beta-citronellol, geraniol, phenylacetic, indolacetic, hexanoic and octanoic acids were tested against resistant strains of the human pathogens, Escherichia coli, Staphylococcus aureus and Candida albicans. Assays were carried out using filter paper discs impregnated with either hexane or water solutions of the analytes in the concentration range 250-6,000 ng/microl. Although most of the tested compounds presented strong antibacterial and antifungal activities, citral, geraniol, 4-methyl-3-heptanol, hexanoic and octanoic acids were the most effective, particularly against C. albicans. The results suggest that these compounds may be of potential value as antibiotics in the treatment of human candidiasis.
Product Description
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
3-Octanone, ≥98%, FG
Sigma-Aldrich
3-Octanone, ≥98%
Sigma-Aldrich
2-Heptanone, 99%
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG