Merck
  • Mass spectrometry-compatible silver staining of histones resolved on acetic acid-urea-Triton PAGE.
Mass spectrometry-compatible silver staining of histones resolved on acetic acid-urea-Triton PAGE.
Proteomics (2009-04-04)
Khare Satyajeet Pramod, Khade Bharat, Gupta Sanjay
ABSTRACT
Acetic acid-Urea-Triton (AUT) PAGE is commonly used method to separate histone variants and their post-translationally modified forms. Coomassie staining is the preferred method for protein visualization; however, its sensitivity is less than that of silver staining. Though silver staining of histones in AUT-PAGE has been reported, the method is time-consuming, dependent on prior staining by Amido black and has not been reported suitable for mass spectrometry. Here, we propose 'SDS-Silver' method for rapid, sensitive and mass spectrometry-compatible staining of histones resolved on AUT-PAGE.
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent