Merck
  • Competitive biodegradation of dichlobenil and atrazine coexisting in soil amended with a char and citrate.
Competitive biodegradation of dichlobenil and atrazine coexisting in soil amended with a char and citrate.
Environmental pollution (Barking, Essex : 1987) (2009-07-01)
Yuping Qiu, Hui Pang, Zunlong Zhou, Ping Zhang, Yucheng Feng, G Daniel Sheng
ABSTRACT
The role of char nutrients in the biodegradation of coexisting dichlobenil and atrazine in a soil by their respective bacterial degraders, DDN and ADP, was evaluated. Under growing conditions, their degradation in soil extract was slow with <40% and <20% degraded within 64 h, respectively. The degradation in extracts and slurries of char-amended solids increased with increasing char content, due to nutritional stimulation on microbial activities. By supplementing soil extract with various major nutrients, the measured degradation demonstrated that P was the exclusive limiting nutrient. The reduction in the degradation of coexisting dichlobenil and atrazine resulted apparently from the competitive utilization of P by DDN and ADP. With a shorter lag phase, ADP commenced growing earlier than DDN with the advantage of utilizing P first in insufficient supply. This resulted in an inhibition on the growth of DDN and thus suppression on dichlobenil degradation.
Sigma-Aldrich
2,6-Dichlorobenzonitrile, 97%