Merck
Hand dermatitis: a focus on allergic contact dermatitis to biocides.
Dermatologic clinics (2009-07-08)
Lisa E Maier, Heather P Lampel, Tina Bhutani, Sharon E Jacob
ABSTRACT
Hand dermatitis is a common disease of the skin resulting in significantly decreased quality of life. Allergic contact dermatitis is a frequent cause of hand dermatitis. Recent studies have revealed that biocides used as preservatives are frequent allergens affecting the hands. This article reviews common biocides implicated in hand dermatitis.
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, 99%
Supelco
Methyldibromoglutaronitrile, analytical standard
Sigma-Aldrich
4-Chloro-3,5-dimethylphenol, purum, ≥98.0% (T)
Sigma-Aldrich
Imidazolidinyl urea
Sigma-Aldrich
3-Iodo-2-propynyl N-butylcarbamate, 97%