Merck
  • Pressure and temperature dependence of the reaction of vinyl radical with alkenes III: measured rates and predicted product distributions for vinyl + butene.
Pressure and temperature dependence of the reaction of vinyl radical with alkenes III: measured rates and predicted product distributions for vinyl + butene.
The journal of physical chemistry. A (2009-07-25)
C Franklin Goldsmith, Huzeifa Ismail, William H Green
ABSTRACT
This work reports experimental and theoretical first-order rate constants for the reaction of vinyl radical with C(4)H(8) alkenes: 1-butene, 2-butene, and isobutene. The experiments are performed over a temperature range of 300 to 700 K at 100 Torr. Vinyl radicals (H(2)C horizontal lineCH) were generated by laser photolysis of vinyl iodide (C(2)H(3)I) at 266 nm, and time-resolved absorption spectroscopy was used to probe vinyl radicals at 423.2 and 475 nm. Weighted Arrhenius fits to the experimental rate coefficients for 1-butene (k(1)), 2-butene (k(2)), and isobutene (k(3)) yield k(1) = (1.3 +/- 0.3) x 10(-12) cm(3) molecules(-1) s(-1) exp[-(2200 +/- 120) K/T]; k(2) = (1.7 +/- 0.3) x 10(-12) cm(3) molecules(-1) s(-1) exp[-(2610 +/- 120) K/T]; and k(3) = (1.0 +/- 0.1) x 10(-12) cm(3) molecules(-1) s(-1) exp[-(2130 +/- 50) K/T], respectively. C(6)H(11) potential energy surfaces (PESs) for each system were calculated using the G3 method. RRKM/ME simulations were performed for each system to predict pressure-dependent rate coefficients and branching fractions for the major channels. A generic rate rule for vinyl addition to various alkenes is recommended; a similar rate rule for the abstraction of H atoms by vinyl from alkenes is also provided. Some of the vinyl addition reactions exhibit anomalous Evans-Polanyi plots similar to those reported for previous methyl addition reactions.
Sigma-Aldrich
1-Butene, ≥99%
Sigma-Aldrich
2-Methylpropene, purum, ≥99.5%
Sigma-Aldrich
2-Methylpropene, 99%