Merck
  • Enhanced production of 6-aminopenicillanic acid in aqueous methyl isobutyl ketone system with immobilized penicillin G acylase.
Enhanced production of 6-aminopenicillanic acid in aqueous methyl isobutyl ketone system with immobilized penicillin G acylase.
Preparative biochemistry & biotechnology (2009-12-22)
Shu-Guang Fang, Tan Qiang, Rui-Jiang Liu, Xi-Ming Xu, Ye-Wang Zhang
ABSTRACT
Enzymatic hydrolysis of penicillin G for production of 6-amino-penicillanic-acid (6-APA) was achieved by using penicillin G acylase as catalyst in an aqueous-methylisobutyl ketone (MIBK) system. The optimization was carried out and it was found that the best conversion was improved 10% more than the aqueous system, which was obtained at the conditions: initial pH 8.0, 5.0% (W/V) substrate (penicillin G), and temperature at 35 degrees C, and the ratio of aqueous and organic phase was 3:1. The stability of the biocatalyst was studied at the operational conditions. After 5 cycles of semi-batch reactions, the residual activity of penicillin G acylase was 69.2% of the initial activity. There was no apparent loss of the yield of product. This process has a potential application in the industrial scale production of 6-APA because it simplifies the process effectively.
Sigma-Aldrich
(+)-6-Aminopenicillanic acid, 96%
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Sigma-Aldrich
2-Hexanone, reagent grade, 98%
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Supelco
4-Methyl-2-pentanone, analytical standard
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Supelco
2-Hexanone, analytical standard
Supelco
2-Hexanone, analytical standard