Merck
  • Forensic analysis of hallucinogenic mushrooms and khat (Catha edulis Forsk) using cation-exchange liquid chromatography.
Forensic analysis of hallucinogenic mushrooms and khat (Catha edulis Forsk) using cation-exchange liquid chromatography.
Forensic science international (2010-01-06)
Tim Laussmann, Sigrid Meier-Giebing
ABSTRACT
Hallucinogenic mushrooms (e.g. Psilocybe and Panaeolus species) as well as leaves and young shoots of the khat tree (Catha edulis Forsk) are illicit drugs in many countries. The exact concentration of the hallucinogenic alkaloids psilocin and psilocybin in mushrooms and the sympathomimetic alkaloids cathinone and cathine in khat is usually essential for jurisdiction. Facing an increasing number of mushroom and khat seizures by German customs authorities, a convenient comprehensive quantitative HPLC method based on cation-exchange liquid chromatography for these rather "exotic" drugs has been developed which avoids time-consuming multi-step sample preparation or chemical derivatization procedures. Using this method a number of different hallucinogenic fungi species and products that are mainly distributed via the internet have been analysed (dried and fresh Psilocybe cubensis Singer as well as P. cubensis collected from "grow boxes", Panaeolus cyanescens Berkeley and Broome and so-called "philosopher stones" (sclerotia of Psilocybe species)). Highest total amounts of psilocin have been detected in dried P. cyanescens reaching up to 3.00+/-0.24 mg per 100 mg. The distribution of khat alkaloids in different parts of the khat shoots has been studied. High concentrations of cathinone have not only been detected in leaves but also in green parts and barks of stalks. Additionally, the sample treatment for fresh mushroom and khat samples has been optimised. Highest amounts of alkaloids were found when fresh material was freeze-dried.
Sigma-Aldrich
DL-Norephedrine hydrochloride, ≥99%