Merck
  • Determination of aflatoxins B1, B2, G1, G2 and ochratoxin A in animal feed by ultra high-performance liquid chromatography-tandem mass spectrometry.
Determination of aflatoxins B1, B2, G1, G2 and ochratoxin A in animal feed by ultra high-performance liquid chromatography-tandem mass spectrometry.
Journal of separation science (2010-02-04)
Sergio José López Grío, Antonia Garrido Frenich, José Luis Martínez Vidal, Roberto Romero-González
ABSTRACT
A rapid and simple method was developed to determine aflatoxins B1, B2, G1, G2 and ochratoxin A in animal feed and pet foods by UHPLC-MS/MS. Because the complexity of the evaluated matrices, the proposed method is based on sonication extraction using an ACN/water mixture (80:20 v/v) followed by a clean-up step utilising C(18) as sorbent. Performance parameters of the method were evaluated, including linearity, trueness, precision and LOQ. Good linearity was found for all mycotoxins, with determination coefficients higher than 0.99 in the range considered, using matrix-matched calibration for quantification purposes. Recoveries ranged from 84 to 113%, with RSD lower than 20%, whereas LOQs were 5 microg/kg for the assayed mycotoxins. Finally, the method was successfully applied to the analysis of 19 real samples, detecting aflatoxin G2 in two samples at 13 and 17 microg/kg respectively, whereas the other mycotoxins were detected at trace levels (<LOQ) in five samples.
Product Description
Aflatoxin G1 solution, 3.78 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin G2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Aflatoxin B2 solution, 3.80 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin G1 solution, 2 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin B2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Supelco
Aflatoxin G2 solution, 0.5 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
Aflatoxin B2
Supelco
Aflatoxin G1
Sigma-Aldrich
Aflatoxin G1, from Aspergillus flavus
Sigma-Aldrich
Aflatoxin G2