Merck
  • The synthesis of novel fluorescent purine analogues modified by azacrown ether at C6.
The synthesis of novel fluorescent purine analogues modified by azacrown ether at C6.
Bioorganic & medicinal chemistry letters (2010-04-20)
Hai-Ming Guo, Jing Wu, Hong-Ying Niu, Dong-Chao Wang, Feng Zhang, Gui-Rong Qu
ABSTRACT
The synthesis and fluorescence properties of novel purine analogues linked azacrown ether at C6 position were investigated. These new purine analogues could be prepared from a series of 6-chloropurines and showed selective and efficient signaling behaviors toward micromolar concentration of Ag(+) ion over other common metal ions in an aqueous environment.
Sigma-Aldrich
6-Chloropurine, ≥99%
Sigma-Aldrich
6-Chloropurine, 99%