Merck
  • Retention of aroma compounds: an interlaboratory study on the effect of the composition of food matrices on thermodynamic parameters in comparison with water.
Retention of aroma compounds: an interlaboratory study on the effect of the composition of food matrices on thermodynamic parameters in comparison with water.
Journal of the science of food and agriculture (2010-05-18)
Mirela Kopjar, Isabelle Andriot, Anne Saint-Eve, Isabelle Souchon, Elisabeth Guichard
ABSTRACT
Partition coefficients give an indication of the retention of aroma compounds by the food matrix. Data in the literature are obtained by various methods, under various conditions and expressed in various units, and it is thus difficult to compare the results. The aim of the present study was first to obtain gas/water and gas/matrix partition coefficients of selected aroma compounds, at different temperatures, in order to calculate thermodynamic parameters and second to compare the retention of these aroma compounds in different food matrices. Yogurts containing lipids and proteins induced a higher retention of aroma compounds than model gel matrices. The observed effects strongly depend on hydrophobicity of aroma compounds showing a retention for ethyl hexanoate and a salting out effect for ethyl acetate. A small but noticeable decrease in enthalpy of affinity is observed for ethyl butyrate and ethyl hexanoate between water and food matrices, suggesting that the energy needed for the volatilization is lower in matrices than in water. The composition and complexity of a food matrix influence gas/matrix partition coefficients or aroma compounds in function of their hydrophobicity and to a lower extent enthalpy of vaporization.
Sigma-Aldrich
Ethyl hexanoate, ≥99%
Sigma-Aldrich
Ethyl hexanoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl butyrate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl hexanoate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl butyrate, ≥98%, FCC, FG
Sigma-Aldrich
Ethyl butyrate, 99%
Supelco
Ethyl butyrate, analytical standard