Merck
  • Determination of five phthalate monoesters in human urine using gas chromatography-mass spectrometry.
Determination of five phthalate monoesters in human urine using gas chromatography-mass spectrometry.
Bulletin of environmental contamination and toxicology (2010-06-25)
Fumio Kondo, Yoshitomo Ikai, Rumiko Hayashi, Masanao Okumura, Satoshi Takatori, Hiroyuki Nakazawa, Shun-ichiro Izumi, Tsunehisa Makino
ABSTRACT
We have developed a gas chromatography-mass spectrometry (GC-MS) method to determine five phthalate monoesters (monoethyl phthalate (MEP), mono-n-butyl phthalate (MBP), mono-(2-ethylhexyl) phthalate (MEHP), monoisononyl phthalate (MINP) and monobenzyl phthalate (MBz)) in human urine. Human urine samples were subjected to enzymatic deconjugation of the glucuronides followed by extraction with hexane. The extracted phthalate monoesters were methylated with diazomethane, purified on a Florisil column and then subjected to GC-MS analysis. The recoveries from urine spiked with five phthalate monoesters were 86.3%-119% with coefficients of variation of 0.6%-6.1%. We measured phthalate monoester levels in human urine by analyzing 36 samples from volunteers. MBP and MEP were detected in all samples, and their median concentrations were 60.0 and 10.7 ng/mL, respectively. MBzP and MEHP were found in 75% and 56% of samples, and their median concentrations were 10.9 and 5.75 ng/mL, respectively. MINPs were not detected in most samples (6% detectable). Women had significantly (p < 0.05) higher mean concentrations of MBP and MEP than men. The estimated daily exposure levels for the four parent phthalates excluding diisononyl phthalate ranged from 0.27 to 5.69 mug/kg/day (median).
Sigma-Aldrich
mono-Butyl phthalate, 97.0-103.0% (T)
Supelco
Benzyl butyl phthalate, analytical standard
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Supelco
Benzyl butyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland