Merck
  • Simultaneous quantification of dextromethorphan and its metabolites dextrorphan, 3-methoxymorphinan and 3-hydroxymorphinan in human plasma by ultra performance liquid chromatography/tandem triple-quadrupole mass spectrometry.
Simultaneous quantification of dextromethorphan and its metabolites dextrorphan, 3-methoxymorphinan and 3-hydroxymorphinan in human plasma by ultra performance liquid chromatography/tandem triple-quadrupole mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2010-10-12)
Walter J Loos, Anne-Joy M de Graan, Peter de Bruijn, Ron H N van Schaik, Marianne A C van Fessem, Mei-Ho Lam, Ron H J Mathijssen, Erik A C Wiemer
ABSTRACT
A rapid and sensitive ultra performance liquid chromatography/tandem mass spectrometry (UPLC-MS/MS) method has been developed and validated for the simultaneous quantitative determination of dextromethorphan (DM) and its metabolites dextrorphan (DX), 3-methoxymorphinan (3MM) and 3-hydroxymorphinan (3HM), in human lithium heparinized plasma. The extraction involved a simple liquid-liquid extraction with 1 ml n-butylchloride from 200μl aliquots of plasma, after the addition of 20 μl 4% (v/v) ammonium hydroxide and 100 μl stable labeled isotopic internal standards in acetonitrile. Chromatographic separations were achieved on an Aquity UPLC(®) BEH C(18) 1.7 μm 2.1 mm x 100mm column eluted at a flow-rate of 0.250 ml/min on a gradient of acetonitrile. The overall cycle time of the method was 7 min, with elution times of 1.3min for DX and 3HM, 2.8 min for 3MM and 2.9min for DM. The multiple reaction monitoring transitions were set at 272>215 (m/z), at 258>133 (m/z), at 258>213 (m/z) and at 244>157 (m/z) for DM, DX, 3MM and 3HM, respectively. The calibration curves were linear (r²≥0.995) over the range of 0.500-100 nM with the lower limit of quantitation validated at 0.500 nM for all compounds, which is equivalent to 136, 129, 129 and 122 pg/ml for DM, DX, 3MM and 3HM, respectively. Extraction recoveries were constant, but ranged from 39% for DM to 83% for DX. The within-run and between-run precisions were within 11.6%, while the accuracy ranged from 92.7 to 110.6%. The applicability of the bioanalytical method was demonstrated and is currently implemented in a clinical trial to study DM as probe-drug for individualized tamoxifen treatment in breast cancer patients.