Merck
Macroglomeruli for fruit odors change blend preference in Drosophila.
Die Naturwissenschaften (2010-10-26)
Irene Ibba, Anna Maria Angioy, Bill S Hansson, Teun Dekker
ABSTRACT
The olfactory circuitry of Drosophila melanogaster is becoming increasingly clear. However, how olfactory processing translates into appropriate behavioral responses is still poorly understood. Using a sibling species approach, we tested how a perturbation in the olfactory circuitry affects odor preference. In a previous study, we found that the sibling species of D. melanogaster, the specialist D. sechellia, overrepresents a sensillum, ab3, the A neuron of which is sensitive to hexanoate esters, characteristic of the species' sole host, the Morinda citrifolia fruit. Concordantly, the corresponding glomerulus, DM2, is enlarged. In this study, we found that the ab3B neuron, the expansion of which was previously assumed to be pleiotropic and of no ecological significance, is in fact tuned to another morinda fruit volatile, 2-heptanone (HP). Axons of this neuron type arborize in a second enlarged glomerulus. In behavioral experiments we tested how this has affected the fly's odor preference. We demonstrate that D. sechellia has a reversed preference for the key ligands of these macroglomeruli, especially at high concentrations. Whereas D. melanogaster was repelled by high concentrations of these odors, D. sechellia was highly attracted. This was the case for odors presented singly, but more notably for blends thereof. Our study indicates that relatively simple changes, such as a shift in sensillar abundance, and concordant shifts in glomerular size, can distort the resulting olfactory code, and can lead to saltatory shifts in odor preference. D. sechellia has exploited this to align its olfactory preference with its ecological niche.
Sigma-Aldrich
Hexanoic acid, purum, ≥98.0% (GC)
Supelco
Methyl hexanoate, analytical standard
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Methyl hexanoate, 99%
Sigma-Aldrich
Methyl hexanoate, natural, ≥99%, FG
Sigma-Aldrich
Methyl hexanoate, ≥99%, FG
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Sodium hexanoate, 99-100%
Supelco
Hexanoic acid, analytical standard
Sigma-Aldrich
2-Heptanone, 99%
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG