Merck
  • Treatment of waste gas from the breather vent of a vertical fixed roof p-xylene storage tank by a trickle-bed air biofilter.
Treatment of waste gas from the breather vent of a vertical fixed roof p-xylene storage tank by a trickle-bed air biofilter.
Bioresource technology (2010-11-03)
Shenteng Chang, Chungsying Lu, Shihchieh Hsu, How-Tsan Lai, Wen-Lin Shang, Yeong-Song Chuang, Chi-Huang Cho, Sheng-Han Chen
ABSTRACT
This study applied a pilot-scale trickle-bed air biofilter (TBAB) system for treating waste gas emitted from the breather vent of a vertical fixed roof storage tank containing p-xylene (p-X) liquid. The volatile organic compound (VOC) concentration of the waste gas was related to ambient temperature as well as solar radiation, peaking at above 6300 ppmv of p-X and 25000 ppmv of total hydrocarbons during the hours of 8 AM to 3 PM. When the activated carbon adsorber was employed as a VOC buffer, the peak waste gas VOC concentration was significantly reduced resulting in a stably and efficiently performing TBAB system. The pressure drop appeared to be low, reflecting that the TBAB system could be employed in the prolonged operation with a low running penalty. These advantages suggest that the TBAB system is a cost-effective treatment technology for VOC emission from a fixed roof storage tank.
Sigma-Aldrich
p-Xylene, anhydrous, ≥99%
Supelco
p-Xylene, analytical standard
Sigma-Aldrich
p-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
p-Xylene, suitable for HPLC, ≥99%
Supelco
p-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material