Merck
Chemical composition of the essential oil of Feronia elephantum Correa.
Natural product research (2010-11-26)
Chitra Pande, Geeta Tewari, Charu Singh, Shalini Singh, R C Padalia
ABSTRACT
The essential oil composition of Feronia elephantum Correa (family: Rutaceae) was examined by capillary gas chromatography (GC) and gas chromatography-mass spectroscopy (GC-MS). The analysis revealed the presence of 24 constituents, of which 18 constituents were identified. Trans-anethole (57.73%) and methyl chavicol (37.48%) were the major compounds, while cis-anethole, p-anisaldehyde, (E)-jasmone, methyl eugenol, β-caryophyllene, linalool and (E)-methyl isoeugenol were also present as the minor constituents.
Product Description
Supelco
p-Anisaldehyde, analytical standard
Sigma-Aldrich
Methyl isoeugenol, ≥98%, FG
Sigma-Aldrich
1,2-Dimethoxy-4-propenylbenzene, 99%
Sigma-Aldrich
p-Anisaldehyde, 98%
Supelco
Estragole, analytical standard
Sigma-Aldrich
p-Anisaldehyde, ≥97.5%, FCC, FG
Sigma-Aldrich
cis-Jasmone, stabilized
Supelco
Jasmone, analytical standard
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
4-Allylanisole, 98%
Sigma-Aldrich
4-Allyl-1,2-dimethoxybenzene, 99%
Sigma-Aldrich
trans-Anethole, 99%
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
4-Allylanisole, ≥98%, FCC
Supelco
Methyl eugenol, analytical standard
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Sigma-Aldrich
Methyl eugenol, ≥98%, FCC