Merck
  • A sub-nanomole level electrochemical method for determination of prochloraz and its metabolites based on medical stone doped disposable electrode.
A sub-nanomole level electrochemical method for determination of prochloraz and its metabolites based on medical stone doped disposable electrode.
Talanta (2010-11-30)
Hongbo Li, Jing Li, Chuantao Hou, Shi Du, Yanyan Ren, Zhanjun Yang, Qin Xu, Xiaoya Hu
ABSTRACT
A ultrasensitive, simple and convenient electrochemical method was firstly developed for the determination of prochloraz and its metabolites as 2,4,6-trichlorophenol (2,4,6-TCP) using nano-aperture medical stone. Compared with the undoped disposable electrode (UDE), nano-aperture medical stone doped disposable electrode (MSDDE) not only significantly enhances the oxidation peak current of 2,4,6-TCP but also lowers the oxidation overpotential, suggesting that the nano-aperture MSDDE can remarkably improve the sensitivity of 2,4,6-TCP. The experimental conditions such as pH values of buffer solution, the content of nano-aperture medical stone, accumulation potential and time were optimized for the determination of 2,4,6-TCP. At optimal conditions, the oxidation peak current is proportional to the concentration of 2,4,6-TCP over the range from 6.0 × 10(-9) to 8.0 × 10(-5)mol L(-1). Finally, this novel method was successfully employed to detect prochloraz and its metabolites in orange rind with the detection limit of 8.4 × 10(-10)mol L(-1) (0.3 ng g(-1)) and the method was validated by gas chromatography.
Product Description
Supelco
Prochloraz, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Prochloraz, PESTANAL®, analytical standard