Merck
  • Inhibitory effects of UV-based therapy on dry skin-inducible nerve growth in acetone-treated mice.
Inhibitory effects of UV-based therapy on dry skin-inducible nerve growth in acetone-treated mice.
Journal of dermatological science (2011-04-05)
Atsuko Kamo, Mitsutoshi Tominaga, Suhandy Tengara, Hideoki Ogawa, Kenji Takamori
ABSTRACT
UV-based therapy has anti-pruritic effects in inflammatory skin diseases, such as atopic dermatitis and psoriasis. These anti-pruritic effects may be partly due to inhibition of intraepidermal nerve growth, but they have not been fully characterized. This study was performed to characterize the anti-nerve growth effects of UV-based therapies in acetone-treated mice as an acute dry skin model. Nerve fibers penetrate into the epidermis 24h after acetone treatment in mice, and nerve growth peaks 48h after acetone treatment. To investigate the effects of UV-based therapies on the epidermal nerve fibers, including combination treatment with corticosteroid ointment, the mice were treated with psoralen ultraviolet A (PUVA), PUVA and betamethasone valerate ointment (PUVA+BV), narrowband ultraviolet B (NB-UVB), or an excimer lamp. Each therapy was provided 24h after acetone treatment, and skin samples were taken 48h later. Nerve fiber densities and expression levels of nerve growth factor (NGF) and semaphorin 3A (Sema3A) in the epidermis were examined by immunohistochemistry. Penetration of nerve fibers into the epidermis was observed in the acetone-treated mice, concomitant with increased NGF and decreased Sema3A levels in the epidermis. The acetone-induced intraepidermal nerve growth was significantly decreased by PUVA, PUVA+BV, NB-UVB, and excimer lamp treatments compared with controls. In addition, PUVA+BV and NB-UVB normalized the abnormal expression of NGF and Sema3A in the epidermis, but no such normalization was observed with excimer lamp treatment. UV-based therapies, especially NB-UVB and excimer lamp treatments, may be effective therapeutic methods for pruritus involving epidermal hyperinnervation.
Product Description
Betamethasone valerate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betamethasone 17-valerate
Supelco
Betamethasone 17-valerate, VETRANAL®, analytical standard
Betamethasone 17-valerate, European Pharmacopoeia (EP) Reference Standard