- Studies on immunoassays of peptide factors. V. Synthesis of cholecystokinin-58-[1-11]/iso-1-cytochrome c conjugate.
Studies on immunoassays of peptide factors. V. Synthesis of cholecystokinin-58-[1-11]/iso-1-cytochrome c conjugate.
Biological chemistry Hoppe-Seyler (1987-07-01)
L Moroder, C Tzougraki, W Göhring, G Mourier, H J Musiol, E Wünsch
PMID3040038
ABSTRACT
In previous studies on model compounds we have found that the maleimide function is sufficiently stable under the conditions of peptide synthesis to allow its incorporation at preselected positions of a peptide chain in earlier steps of the synthetic route. Taking advantage of this observation the N-terminal undecapeptide of canine cholecystokinin-58 containing at its N-terminus the maleimide group became accessible in high yields as chromatographically homogenous and analytically well characterized compound. Via the incorporated anchor group the undecapeptide was linked selectively at its N-terminus to the cysteine residue 107 of iso-1-cytochrome c to yield a well characterized conjugate of 1:1 stoichiometry for immunization experiments.