Merck
  • Synthesis of buprenorphine from oripavine via N-demethylation of oripavine quaternary salts.
Synthesis of buprenorphine from oripavine via N-demethylation of oripavine quaternary salts.
The Journal of organic chemistry (2011-04-19)
Lukas Werner, Ales Machara, David R Adams, D Phillip Cox, Tomas Hudlicky
ABSTRACT
Buprenorphine was synthesized from oripavine by a sequence involving the conversion of oripavine into its cyclopropylmethyl quaternary salt, N-demethylation with thiolate to N-cyclopropylmethyl nororipavine, and conversion of this material to the title compound by previously available methods. The new synthesis avoids toxic reagents used previously, is shorter, and proceeds in comparable yields. Experimental and spectral data are provided for all new compounds.
Sigma-Aldrich
Cyanogen bromide, 99.995% trace metals basis
Sigma-Aldrich
Cyanogen bromide, ≥98.5% (RT)
Sigma-Aldrich
Cyanogen bromide, reagent grade, 97%