Merck
  • Embryopathic effects of thalidomide and its hydrolysis products in rabbit embryo culture: evidence for a prostaglandin H synthase (PHS)-dependent, reactive oxygen species (ROS)-mediated mechanism.
Embryopathic effects of thalidomide and its hydrolysis products in rabbit embryo culture: evidence for a prostaglandin H synthase (PHS)-dependent, reactive oxygen species (ROS)-mediated mechanism.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2011-04-20)
Crystal J J Lee, Luisa L Gonçalves, Peter G Wells
ABSTRACT
Thalidomide (TD) causes birth defects in humans and rabbits via several potential mechanisms, including bioactivation by embryonic prostaglandin H synthase (PHS) enzymes to a reactive intermediate that enhances reactive oxygen species (ROS) formation. We show herein that TD in rabbit embryo culture produces relevant embryopathies, including decreases in head/brain development by 28% and limb bud growth by 71% (P<0.05). Two TD hydrolysis products, 2-phthalimidoglutaramic acid (PGMA) and 2-phthalimidoglutaric acid (PGA), were similarly embryopathic, attenuating otic vesicle (ear) and limb bud formation by up to 36 and 77%, respectively (P<0.05). TD, PGMA, and PGA all increased embryonic DNA oxidation measured as 8-oxoguanine (8-oxoG) by up to 2-fold (P<0.05). Co- or pretreatment with the PHS inhibitors eicosatetraynoic acid (ETYA) or acetylsalicylic acid (ASA), or the free-radical spin trap phenylbutylnitrone (PBN), completely blocked embryonic 8-oxoG formation and/or embryopathies initiated by TD, PGMA, and PGA. This is the first demonstration of limb bud embryopathies initiated by TD, as well as its hydrolysis products, in a mammalian embryo culture model of a species susceptible to TD in vivo, indicating that all likely contribute to TD teratogenicity in vivo, in part through PHS-dependent, ROS-mediated mechanisms.
Sigma-Aldrich
5,8,11,14-Eicosatetraynoic acid, ≥97%