Merck
  • Absorption of levothyroxine when coadministered with various calcium formulations.
Absorption of levothyroxine when coadministered with various calcium formulations.
Thyroid : official journal of the American Thyroid Association (2011-05-21)
Isabelle Zamfirescu, Harold E Carlson
ABSTRACT
Calcium carbonate is a commonly used dietary supplement and has been shown to interfere with levothyroxine absorption. However, calcium citrate, which is also used for supplementation purposes, has not been studied previously and calcium acetate, which is used to treat hyperphosphatemia in renal failure, has been reported to show little or no interference with levothyroxine absorption in a retrospective pharmacoepidemiologic study. We aimed to compare the effect of these three calcium formulations on levothyroxine absorption. The study was conducted in eight healthy, euthyroid adults. We performed single-dose pharmacokinetic studies in which we measured levothyroxine absorption when given alone or when coadministered with calcium carbonate, calcium citrate, or calcium acetate in doses containing 500 mg elemental calcium. Serum thyroxine was measured at intervals over a 6-hour period after ingestion of the study drugs. Coadministration of each of the three calcium preparations significantly reduced levothyroxine absorption by about 20%-25% compared with levothyroxine given alone. Contrary to a prior report, our data suggest that calcium acetate interferes with levothyroxine absorption in a manner similar to that seen with calcium carbonate and calcium citrate. Although the effect of calcium is modest compared with some other medications previously studied, hypothyroid patients should be cautioned to take their levothyroxine well-separated from all of these calcium formulations.
Product Description
Supelco
Calcium Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium citrate tetrahydrate, 99%