Merck
  • [Determination of trimethyltin chloride in urine by headspace-gas chromatography].
[Determination of trimethyltin chloride in urine by headspace-gas chromatography].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2011-05-31)
Zheng Ruan, Hong-fang Tang, Dan-hua Liu, Cheng-min Xu, Ya-Ling Qian
ABSTRACT
To establish a detection method for trimethyltin chloride in urine by the Head space-GC. After derivatizing trimethyltin chloride, the urines was separated by the head space-gc, and then the trimethyltin chloride detected qualitatively and quantificationally. In the concentration range of 0.02 ∼ 0.40 mg/L urinary trimethyltin chloride, showed a quadratic, r = 0.9992, detection limit was 0.005 mg/L, the relative standard deviation was 1.9% ∼ 2.5%, recovery was 92.0% to 100%, the urine samples can be saved at least 90 days in -18°C refrigerator. The instrument, reagents involved in the detection require low, the operations to processing samples are simple, high sensitivity, less interference, good reproducibility, and suitable for quantitative and qualitative analysis, convenient to promotion.
Sigma-Aldrich
Trimethyltin chloride
Sigma-Aldrich
Trimethyltin chloride solution, 1.0 M in THF
Sigma-Aldrich
Trimethyltin chloride solution, 1.0 M in hexanes