Merck
  • PVC membrane sensor for potentiometric determination of iron (II) in some pharmaceutical formulations based on a new neutral ionophore.
PVC membrane sensor for potentiometric determination of iron (II) in some pharmaceutical formulations based on a new neutral ionophore.
Drug testing and analysis (2011-06-24)
M A Abounassif, M A Al-Omar, A-G E Amr, G A E Mostafa
ABSTRACT
A novel poly (vinyl chloride) PVC membrane sensor for Fe(2+) ions is described. The sensor is based on the use of newly synthesized chiral 2,6-bis-(carboxamide methyl ester)pyridine derivative as neutral ionophore in plasticized PVC membrane. The sensor display a fast, stable and near-Nernstian response over a relative wide ferrous concentration range (1 × 10(-3) to 6 × 10(-6) M), with cationic slope of 31.5 ± 0.5, mV per concentration decade over a pH range of 5.0-9.0. The direct determination of 0.25-56.0 µg/ml of ferrous in aqueous solution shows an average recovery of 98.5% and a mean relative standard deviation of 1.5% at 20.0 µg/ml. The sensor displays long life-span, long-term stability, high reproducibility, and short response time. Selectivity coefficients for Fe(II) relative to a number of interfering substances were investigated. The sensor shows high significantly for Fe(2+) over Fe,(3+) Cu,(2+) Zn,(2+) Cd,(2+) Hg,(2+) Pb,(2+) Ni,(2+) Co,(2+) Mn,(2+) Al,(3+) alkaline earth and alkali metal ions. The sensor is successfully applied for measurement of ferrous in drug formulations. The results obtained for the determination of ferrous using the proposed sensor are comparable favourably with those obtained using the spectrophotometric method. Copyright © 2010 John Wiley & Sons, Ltd.
Sigma-Aldrich
Dibutyl sebacate, ≥97.0% (GC)
Sigma-Aldrich
Dibutyl sebacate, technical grade
Supelco
Dibutyl sebacate, Pharmaceutical Secondary Standard; Certified Reference Material