Merck
  • Biochemical control of bone loss and stone-forming propensity by potassium-calcium citrate after bariatric surgery.
Biochemical control of bone loss and stone-forming propensity by potassium-calcium citrate after bariatric surgery.
Surgery for obesity and related diseases : official journal of the American Society for Bariatric Surgery (2011-06-28)
Khashayar Sakhaee, Carolyn Griffith, Charles Y C Pak
ABSTRACT
Patients undergoing Roux-en-Y gastric bypass (RYGB) surgery are prone to developing bone loss and kidney stones. The goal of the present study was to test the hypothesis that an effervescent formulation of potassium calcium citrate (PCC) would avert metabolic complications by providing bioavailable calcium and alkali. A total of 24 patients with RYGB underwent a 2-phase crossover randomized trial comparing PCC and placebo. During the last 2 days of each 2-week phase, the serum and 24-hour urine samples were analyzed for calcium and bone turnover markers, acid base status, and urinary stone risk factors. Compared with placebo, PCC marginally reduced the serum parathyroid hormone level and significantly decreased urinary deoxypyridinoline by 12% (P <.001) and serum type 1 collagen C-telopeptide by 22% (P <.01). PCC significantly increased the net gastrointestinal alkali absorption, citrate, and pH and significantly lowered the urinary net acid excretion (P <.001). The urinary saturation of uric acid decreased significantly (P <.001). The supersaturation of calcium oxalate and brushite did not change despite an increase in calcium and pH. In untreated urine samples with citrate concentrations altered to mimic those of placebo and PCC, calcium oxalate agglomeration was significantly inhibited by PCC. In RYGB patients, PCC supplementation inhibited bone resorption by providing bioavailable calcium, reduced the urinary saturation of uric acid, and increased the inhibitor activity against calcium oxalate agglomeration by providing alkali that increased urinary pH and citrate.
Sigma-Aldrich
Calcium citrate tetrahydrate, 99%
Sigma-Aldrich
Potassium citrate tribasic monohydrate, ≥98% (GC/titration)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets analytical specification of Ph. Eur., BP, FCC, E332, 99-100.5% (calc with ref. to anhyd. subst.)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Potassium citrate tribasic monohydrate, meets USP testing specifications
Sigma-Aldrich
Potassium citrate tribasic monohydrate, suitable for cell culture