Merck
  • The synthesis and photophysics of tris-heteroleptic cyclometalated iridium complexes.
The synthesis and photophysics of tris-heteroleptic cyclometalated iridium complexes.
Dalton transactions (Cambridge, England : 2003) (2011-07-08)
Robert M Edkins, Alisdair Wriglesworth, Katharina Fucke, Sylvia L Bettington, Andrew Beeby
ABSTRACT
Herein we report the synthesis and photophysical study of tris-heteroleptic complexes of the general formula IrLL'(acac), where L and L' are two differently substituted 2-phenylpyridines (ppyH) and acacH is 2,4-pentanedione, using a combinatorial approach that could be employed for many ligand combinations. The tris-heteroleptic complexes and the analogous bis-heteroleptic complexes of the form IrL(2)(acac) have been studied by a combination of absorption and photoluminescence spectroscopies in conjunction with modelling by DFT and TD-DFT to elucidate the nature and location of the excited state in the novel species.
Sigma-Aldrich
2-Phenylpyridine, 98%
Sigma-Aldrich
Acetylacetone, Wacker Chemie AG, ≥99.5% (GC)
Supelco
Acetylacetone, analytical standard
Sigma-Aldrich
Acetylacetone, ReagentPlus®, ≥99%