Merck
  • [Metabolism inhibition stimulates, metabolism activation inhibits cancerogenic activity of ortho-aminoazotoluene in mouse liver].
[Metabolism inhibition stimulates, metabolism activation inhibits cancerogenic activity of ortho-aminoazotoluene in mouse liver].
Voprosy onkologii (2011-08-04)
V I Kaledin, S I Il'nitskaia
ABSTRACT
Pentachlorophenol, an inhibitor of metabolic activation of aminoazo dyes was administered to suckling mice prior to o-aminoazotoluene (OAT). It was followed by formation of numerous preneoplastic nodules and tumors in the lungs and liver. At the same time, 2,3,7,8-tetrachlorodibenzo-p-dioxine treatment decreased their number in the liver while slightly increasing them in the lung. A possible mechanism of aminoazo dye carcinogenicity is suggested.
Sigma-Aldrich
Fast Garnet GBC base, 97%
Supelco
o-Aminoazotoluene, analytical standard