Merck
Antibacterial potential of citral derivatives.
Natural product communications (2011-09-29)
Soni A Singh, Yogesh A Potdar, Rasika S Pawar, Sujata V Bhat
ABSTRACT
Nine conjugated acid derivatives from monoterpene citral have been synthesized using ester, amide condensations, Wittig, Knoevenagel, Baylis-Hillman reactions and their antimicrobial activities were evaluated through disc diffusion and serial dilution methods. Several derivatives displayed interesting antimicrobial activity. Particularly, the Baylis-Hillman adducts 8-10 were found to have more potent antibacterial and antifungal activities than standard drugs namely chloramphenicol and nystatin respectively.
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Sigma-Aldrich
Citral, 95%