Merck
  • Antimicrobial and antifungal effects of tissue conditioners containing a photocatalyst.
Antimicrobial and antifungal effects of tissue conditioners containing a photocatalyst.
Dental materials journal (2011-09-29)
Masayuki Uchimaru, Takako Sakai, Ryoji Moroi, Susumu Shiota, Yukie Shibata, Mikito Deguchi, Hidetaka Sakai, Yoshihisa Yamashita, Yoshihiro Terada
ABSTRACT
This study examined the antimicrobial/antifungal ability of a tissue conditioner containing a photocatalyst for Escherichia coli, Streptococcus mutans, Staphylococcus aureus and Candida albicans. The photocatalyst was mixed with tissue conditioners powders at concentrations of 0, 10, 15, and 20 wt%. Tissue conditioners powders containing a photocatalyst were mixed with liquid to make test specimens. Test specimens inoculated by each microorganism were irradiated by ultraviolet light for 0-, 2- and 4 hours. The antimicrobial/antifungal effects were evaluated by the CFU technique. The CFU values of each microorganism for tissue conditioners containing a photocatalyst showed significant decrease following UV-irradiation. The improvement in antimicrobial/antifungal effects was concomitant with the increase of the mixing ratio and the irradiation time. Therefore, the results indicated that tissue conditioners containing a photocatalyst might have photocatalytic ability.
Sigma-Aldrich
Poly(ethyl methacrylate), average Mw ~515,000 by GPC, powder
Sigma-Aldrich
Dibutyl sebacate, ≥97.0% (GC)
Sigma-Aldrich
Dibutyl sebacate, technical grade
Supelco
Dibutyl sebacate, Pharmaceutical Secondary Standard; Certified Reference Material