Merck
  • Synthesis and herbicidal activity of diphenyl ether derivatives containing unsaturated carboxylates.
Synthesis and herbicidal activity of diphenyl ether derivatives containing unsaturated carboxylates.
Journal of agricultural and food chemistry (2011-10-01)
Haibo Yu, Huibin Yang, Dongliang Cui, Liang Lv, Bin Li
ABSTRACT
A series of novel diphenyl ether derivatives containing unsaturated carboxylates were designed and synthesized. Their structures were identified by (1)H nuclear magnetic resonance and elemental analyses. The bioassays indicated that the compounds 5b and 5c exhibited good herbicidal activities against velvetleaf at a concentration of 30-40 g/hm(2). The relationship between structure and herbicidal activity was also discussed. Among unsaturated carboxylates group, butenoate is the most promising one. Amonst them, 4-ethoxy-4-oxobutenyl 5-(2-chloro-4-(trifluoromethyl)phenoxy)-2-nitrobenzoate 5b was identified as the most promising candidate due to its high protoporphyrinogen IX oxidase inhibition effect (pI(50) = 6.64) and good herbicidal activity against broadleaf weeds with selectivity to soybean and low toxicity to mammals.
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Supelco
Diphenyl ether, Selectophore, ≥99.9%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG