Merck
  • Visual observation of the mercury-stimulated peroxidase mimetic activity of gold nanoparticles.
Visual observation of the mercury-stimulated peroxidase mimetic activity of gold nanoparticles.
Chemical communications (Cambridge, England) (2011-10-07)
Yi Juan Long, Yuan Fang Li, Yue Liu, Jia Jia Zheng, Jie Tang, Cheng Zhi Huang
ABSTRACT
Mercury-stimulated peroxidase mimetic activity of gold nanoparticles was presented, with which a sensitive label-free colorimetric method for Hg(2+) was developed.
Sigma-Aldrich
Benzidine, ≥98%
Sigma-Aldrich
Benzidine dihydrochloride, ≥99% (titration)
Supelco
Benzidine, analytical standard