Merck
  • VO(acac)(2)/H(2)O(2)/NaI: a mild and efficient combination for the cleavage of dithioacetal derivatives of sugars.
VO(acac)(2)/H(2)O(2)/NaI: a mild and efficient combination for the cleavage of dithioacetal derivatives of sugars.
Carbohydrate research (2011-10-11)
Abu T Khan, Shahzad Ali, R Sidick Basha, Md Musawwer Khan, Mohan Lal
ABSTRACT
A wide variety of dithioacetal derivatives of sugars can be cleaved easily into the corresponding open-chain aldehydo sugars using an efficient combination of VO(acac)(2)/H(2)O(2)/NaI at 0-5°C. Some of the salient features of this protocol are mild reaction conditions, good yields, short reaction times, easy work-up procedures, and non-involvement of toxic chemicals.
Sigma-Aldrich
Copper(II) acetylacetonate, ≥99.9% trace metals basis
Sigma-Aldrich
Manganese(III) acetylacetonate, technical grade
Sigma-Aldrich
Copper(II) acetylacetonate, 97%
Sigma-Aldrich
Sodium iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, 99.999% trace metals basis
Sigma-Aldrich
Sodium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium iodide, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium iodide, ReagentPlus®, ≥99%
Sigma-Aldrich
Chromium(III) acetylacetonate, 97%
Sigma-Aldrich
Manganese(II) acetylacetonate