Merck
  • Chromatographic approaches for determination of low-molecular mass aldehydes in bio-oil.
Chromatographic approaches for determination of low-molecular mass aldehydes in bio-oil.
Journal of chromatography. A (2011-12-14)
Catherine Tessini, Niels Müller, Claudia Mardones, Dietrich Meier, Alex Berg, Dietrich von Baer
ABSTRACT
HPLC-UV and GC/MS determination of aldehydes in bio-oil were evaluated. HPLC-UV preceded by derivatization with 2,4-dinitrophenylhydrazine allows separation and detection of bio-oil aldehydes, but the derivatization affected the bio-oil stability reducing their quantitative applicability. GC/MS determination of aldehydes was reached by derivatization with o-(2,3,4,5,6-pentafluorobenzyl) hydroxylamine hydrochloride. Two approaches for this reaction were evaluated. The first: "in solution derivatization and head space extraction" and the second: "on fiber derivatization SPME", the latter through an automatic procedure. Both sample treatments allows the quantification of most important aliphatic aldehydes in bio-oil, being the SPME approach more efficient. The aldehyde concentrations in bio-oil were ~2% formaldehyde, ~!0.1% acetaldehyde and ~0.05% propionaldehyde.
Product Description
Supelco
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, derivatization grade (GC derivatization), LiChropur, ≥99.0% (AT)
Sigma-Aldrich
O-(2,3,4,5,6-Pentafluorobenzyl)hydroxylamine hydrochloride, ≥98%