Merck
  • Performance of UASB reactor in the biotreatment of 1,1,2-Trichloroethane.
Performance of UASB reactor in the biotreatment of 1,1,2-Trichloroethane.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2012-01-17)
Debolina Basu, Shyam R Asolekar
ABSTRACT
This article aims to examine the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor operated under varying operating conditions, during the treatment of simulated wastewater containing 1,1,2-Trichloroethane (TCA). Initially, reactor R1 (control) and reactor R2 (test) containing TCA were operated at 5 different Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and 12 hours. TCA removal decreased from 99.8% to 96.5%, when the HRT was reduced from 36 to 12 hours in discrete steps. Later, when the OLR was varied periodically from 1.5 to 3.1 kg COD/m(3).d to obtain a different substrate: co-substrate ratios; lower TCA removal efficiencies were observed at an OLR of 2.5 kg COD/ m(3).d and above. The average effluent concentrations of TCA and COD were 0.12 and 79 mg/L, respectively, at the optimum HRT of 24 hours; whereas the effluent TCA and COD concentrations at the substrate: co-substrate ratio of 50:1 was 0.06 mg/L and 48 mg/L, respectively.
Product Description
Supelco
1,1,2-Trichloroethane, analytical standard
Sigma-Aldrich
1,1,2-Trichloroethane, contains ≤3% 2-propanol as stabilizer, 97%