Merck
  • Synergy between prochloraz and esfenvalerate in Daphnia magna from acute and subchronic exposures in the laboratory and microcosms.
Synergy between prochloraz and esfenvalerate in Daphnia magna from acute and subchronic exposures in the laboratory and microcosms.
Aquatic toxicology (Amsterdam, Netherlands) (2012-01-17)
Maj-Britt A Bjergager, Mark L Hanson, Keith R Solomon, Nina Cedergreen
ABSTRACT
Azole fungicides can enhance the toxicity of pyrethroid insecticides on aquatic species. It is, however, unknown to what extend the synergy found under laboratory conditions for strongly sorbing compounds (Azole logK(ow): 3-4, pyrethroid logK(ow): 6-7) will also take place in the field. We therefore investigated the synergising potential of the fungicide prochloraz on the pyrethroid esfenvalerate on Daphnia magna in the laboratory and in situ in cages placed in outdoor aquatic microcosms treated with 0.17, 0.33 and 0.83 μgL(-1) esfenvalerate with and without 90 μg L(-1) prochloraz. We found that the 8-14 fold synergy observed after 2 and 7 days of exposure to mixtures in the microcosms was equivalent to or greater than the 3-7 fold synergy found in 2-day laboratory tests. Incubating new neonates in situ 7 and 14 days after pesticide addition gave EC(50) values of 0.012 ± 0.001 and <0.005μgesfenvalerateL(-1) in the mixture treatments, based on measured water column concentrations. The detection limit is more than ten-fold lower than the lowest esfenvalerate concentration observed to cause ecologically significant effects across seven long term mesocosms studies, hence, even on a longer time scale prochloraz apparently synergises the effect of esfenvalerate under field-like conditions in the microcosms. The results show that synergy found in the laboratory also takes place under field like conditions at quantitatively similar levels, and that it lasts for several weeks. More knowledge on the identification of potential synergists, their true bioavailability and the concentrations and time span within which they can cause synergy needs further study, before an overall evaluation of the occurrence and severity of synergy under field conditions can take place.
Sigma-Aldrich
Fenvalerate, ≥97%
Supelco
Prochloraz, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Prochloraz, PESTANAL®, analytical standard
Supelco
Fenvalerate, PESTANAL®, analytical standard
Supelco
Esfenvalerate, PESTANAL®, analytical standard