Merck
  • Magnetic resonance imaging of the temporomandibular joint disc: feasibility of novel quantitative magnetic resonance evaluation using histologic and biomechanical reference standards.
Magnetic resonance imaging of the temporomandibular joint disc: feasibility of novel quantitative magnetic resonance evaluation using histologic and biomechanical reference standards.
Journal of orofacial pain (2012-01-17)
Hatice T Sanal, Won C Bae, Chantal Pauli, Jiang Du, Sheronda Statum, Richard Znamirowski, Robert L Sah, Christine B Chung
ABSTRACT
To use the ultrashort time-to-echo magnetic resonance imaging (UTE MRI) technique to quantify short T2* properties (obtained through gradient echo) of a disc from the human temporomandibular joint (TMJ) and to corroborate regional T2* values with biomechanical properties and histologic appearance of the discal tissues. A cadaveric human TMJ was sliced sagittally and imaged by conventional and UTE MRI techniques. The slices were then subjected to either biomechanical indentation testing or histologic evaluation, and linear regression was used for comparison to T2* maps obtained from UTE MRI data. Feasibility of in vivo UTE MRI was assessed in two human volunteers. The UTE MRI technique of the specimens provided images of the TMJ disc with greater signal-to-noise ratio (~3 fold) and contrast against surrounding tissues than conventional techniques. Higher T2* values correlated with lower indentation stiffness (softer) and less collagen organization as indicated by polarized light microscopy. T2* values were also obtained from the volunteers. UTE MRI facilitates quantitative characterization of TMJ discs, which may reflect structural and functional properties related to TMJ dysfunction.
Sigma-Aldrich
Direct Red 80, Dye content 25 %
Sigma-Aldrich
Schaeffer and Fulton Spore Stain Solution B
Sigma-Aldrich
Gram′s safranin solution, suitable for microscopy
Sigma-Aldrich
Safranin O, certified by the BSC
Sigma-Aldrich
Safranin O, Dye content ≥85 %
Sigma-Aldrich
Safranin O, suitable for microscopy