Merck
  • Adhesive performance of silver-palladium-copper-gold alloy and component metals bonded with organic sulfur-based priming agents and a tri-n-butylborane initiated luting material.
Adhesive performance of silver-palladium-copper-gold alloy and component metals bonded with organic sulfur-based priming agents and a tri-n-butylborane initiated luting material.
Acta odontologica Scandinavica (2012-02-11)
Miyuki Yamashita, Hiroyasu Koizumi, Takaya Ishii, Daisuke Nakayama, Yusuke Oba, Hideo Matsumura
ABSTRACT
The purpose of the current study was to evaluate the effect of thione-based metal priming agents on the adhesive behavior of a Ag-Pd-Cu-Au alloy and component metals bonded with an acrylic resin. Disk specimens (10 mm in diameter by 3 mm thick) were prepared from a silver-palladium-copper-gold (Ag-Pd-Cu-Au) alloy (Castwell M.C.12), high-purity silver, palladium, copper and gold. Four single-liquid priming agents containing organic sulfur compound (Alloy Primer, Metaltite, M.L. Primer and V-Primer) and three acidic priming agents (All Bond II Primer B, Estenia Opaque Primer and Super-Bond Liquid) were assessed. The metal specimens were flat-ground with abrasive papers, primed with one of the agents and bonded with a tri-n-butylborane initiated resin. The shear bond strengths were determined both before and after repeated thermocycling (5°C and 55°C, 1 min each, 20,000 cycles). The results were statistically analyzed with a non-parametric procedure (p = 0.05 level). The post-thermocycling bond strengths in MPa (median; n = 11) associated with the Alloy Primer, Metaltite, M.L. Primer and V-Primer materials were, respectively, 20.8, 22.8, 17.8 and 18.4 for the Ag-Pd-Cu-Au alloy; 19.6, 21.9, 14.4 and 20.1 for silver; 5.4, 4.5, 12.8 and 5.3 for palladium; 17.1, 19.2, 0.7 and 6.6 for copper; and 18.5, 17.7, 22.8 and 15.4 for gold. It can be concluded that the use of the four priming agents, which are based on organic sulfur compounds, effectively enhanced bonding to the Ag-Pd-Cu-Au alloy and the component metals, although the bonding performance varied among the priming agents and metal elements. The priming agents appeared to have more of an effect on the alloy, silver and gold than on the palladium and copper.
Sigma-Aldrich
2-Thiouracil, ≥99%