Merck
Fragrance material review on benzyl acetate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-03-06)
D McGinty, D Vitale, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of benzyl acetate when used as a fragrance ingredient is presented. Benzyl acetate is a member of the fragrance structural group aryl alkyl alcohol simple acid esters (AAASAE). The AAASAE fragrance ingredients are prepared by reacting an aryl alkyl alcohol with a simple carboxylic acid (a chain of 1-4 carbons) to generate formate, acetate, propionate, butyrate, isobutyrate and carbonate esters. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. Available data for benzyl acetate were evaluated, then summarized, and includes: physical properties, acute toxicity, skin irritation, mucous membrane (eye) irritation, skin sensitization, elicitation, phototoxicity, toxicokinetics, repeated dose, reproductive toxicity, genotoxicity, or carcinogenicity data. A safety assessment of the entire AAASAE will be published simultaneously with this document. Refer Belsito et al. (2012) for an overall assessment of the safe use of this material and all AAASAE in fragrances.
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG
Supelco
Benzyl acetate, analytical standard
Sigma-Aldrich
Benzyl acetate, ≥99%