Merck
  • Understanding the relationship between photolysis efficiency and metal binding using ArgenCast photocages.
Understanding the relationship between photolysis efficiency and metal binding using ArgenCast photocages.
Photochemistry and photobiology (2012-03-13)
Hannah W Mbatia, Daniel P Kennedy, Shawn C Burdette
ABSTRACT
ArgenCast-1 (1), a photocage for silver utilizing acyclic polythioether 3,6,12,15-tetrathia-9-azaheptadecane receptor and 4,5-dimethoxy-2-nitrobenzyl (DMNB) chromophore has been prepared using trimethylsilyl trifluoromethanesulfonate-assisted electrophilic aromatic substitution. Metal binding studies with ArgenCast-1 reveal interactions with Ag(+), Hg(2+) and Cu(+), but only Ag(+) coordinates in both aqueous and organic solvents. The uncaging mechanism of ArgenCast-1 metal complex involves a photoreaction that converts the nitrobenzydrol into the electron withdrawing nitrosobenzophenone that participates in a resonance interaction with a metal-bound aniline nitrogen atom. The structural change following photolysis decreases availability of the nitrogen lone pair for Ag(+) coordination, but strong interactions between Ag(+) and the thioether ligands mitigates metal ion release. A resonance interaction with a key aci-nitro intermediate reduces the photolysis quantum yield of ArgenCast-1, so several naphthyl-based nitrobenzyl groups were screened as alternatives to DMNB. The naphthyl Ag(+) photocages, ArgenCast-2 and -3, exhibit nearly identical quantum yield to ArgenCast-1 owing to the dominance of resonance between aci-nitro intermediate and the aniline nitrogen atom.
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, purum, ≥98.0% (T)
Sigma-Aldrich
Trimethylsilyl trifluoromethanesulfonate, 99%