Merck
  • Potassium acetate and potassium lactate enhance the microbiological and physical properties of marinated catfish fillets.
Potassium acetate and potassium lactate enhance the microbiological and physical properties of marinated catfish fillets.
Journal of food science (2012-03-16)
Sovann Kin, M Wes Schilling, Brian S Smith, Juan L Silva, Taejo Kim, Alessandra J Pham, Stephen G Campano
ABSTRACT
Sodium or potassium salts such as lactate and acetate can be used to inhibit the growth of spoilage bacteria and food-borne pathogens, and thereby prolong the shelf-life of refrigerated seafood. However, minimal information is available regarding the combined effects of potassium salts (acetate and lactate) with an agglomerated phosphate blend on the quality and safety of refrigerated catfish fillets. The objective of this study was to determine the microbiological and quality characteristics of marinated catfish fillets treated with organic acid salts. Catfish fillets were vacuum-tumbled with a brine solution with and without the added organic acid salts, at 10% over initial, raw weight prior to tray-packing and storage at 4 °C for 14 d. Fillets were evaluated for yields, color, pH, tenderness, consumer acceptability, and shelf-life. No differences (P > 0.05) existed among the treated and untreated fillets with regards to solution pick-up and pH, but all treated fillets increased (P < 0.05) cooking yields and Intl. Commission on Illumination (CIE) a* values, and decreased (P < 0.05) CIE L* and b* values in the catfish fillets when compared to the untreated fillets. The fillets treated with a combination of potassium acetate and potassium lactate had lower (P < 0.05) psychrotrophic plate counts and lower spoilage scores than the control treatments on days 7, 10, and 14. In addition, consumers preferred (P < 0.05) treated catfish fillets (fried) with respect to appearance, flavor, and overall acceptability over the negative control. In conclusion, the combination of potassium acetate and potassium lactate enhanced sensory quality and extended the shelf-life of refrigerated catfish fillets.
Sigma-Aldrich
Potassium acetate solution, BioUltra, Molecular Biology, 5 M in H2O
Sigma-Aldrich
Potassium acetate, 99.98% trace metals basis
Sigma-Aldrich
Potassium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium acetate, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium acetate, meets USP testing specifications
Sigma-Aldrich
Potassium acetate, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Potassium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium acetate, ReagentPlus®, ≥99.0%