Merck
  • BF₃·Et₂O-induced stereoselective aldol reaction with benzaldehyde, and steroid sapogenins and its application to a convenient synthesis of dinorcholanic lactones.
BF₃·Et₂O-induced stereoselective aldol reaction with benzaldehyde, and steroid sapogenins and its application to a convenient synthesis of dinorcholanic lactones.
Steroids (2012-03-28)
Karen M Ruíz-Pérez, Margarita Romero-Ávila, Verónica Tinajero-Delgado, Marcos Flores-Álamo, Martín A Iglesias-Arteaga
ABSTRACT
Treatment of steroid sapogenins with benzaldehyde and BF(3)·Et(2)O cleanly produces E-23(23')-benzylidenspirostanes in good yields in a reaction pathway which consists on an aldol reaction followed by a dehydration step. The obtained E-23(23')-benzylidenspirostanes can be easily converted to dinorcholanic lactones by treatment with CrO(3) in acetic acid. The synthetic sequence to dinorcholanic lactones is compatible with the presence of double bonds and carbonyl groups in the steroid framework.
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis
Sigma-Aldrich
Boron trifluoride diethyl etherate
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material