Merck
  • Pubertal effects of 17α-methyltestosterone on GH-IGF-related genes of the hypothalamic-pituitary-liver-gonadal axis and other biological parameters in male, female and sex-reversed Nile tilapia.
Pubertal effects of 17α-methyltestosterone on GH-IGF-related genes of the hypothalamic-pituitary-liver-gonadal axis and other biological parameters in male, female and sex-reversed Nile tilapia.
General and comparative endocrinology (2012-04-07)
Nonglak Phumyu, Surintorn Boonanuntanasarn, Araya Jangprai, Goro Yoshizaki, Uthairat Na-Nakorn
ABSTRACT
The influence of 17α-methyltestosterone (MT) on growth responses, biological parameters and the expression of genes involved in the GH-IGF pathway of the hypothalamic-pituitary-liver-gonadal axis were investigated in female, male, and sex-reversed Nile tilapia to evaluate the relationship between sex and MT-induced changes in these parameters. Female fish had a lower growth rate than male and sex-reversed fish, and MT increased growth performance and duodenal villi in females. Most but not all biological parameters of sex-reversed fish were similar to those of male fish. Male fish had higher red blood cell counts and hemoglobin levels than female and sex-reversed fish, suggesting that these hematological indices reflect a higher metabolic rate in male fish. Greater blood triglyceride levels indicated the vitellogenin process in female fish. MT increased the alternative complement activity in female fish (P<0.05). Sex and MT had no significant effects on the hypothalamic mRNAs of GHRH and PACAP. Although not statistically significant, females tended to have higher GH mRNA levels than male and sex-reversed fish. Additionally, MT tended to decrease and increase GH mRNA levels in female and male fish, respectively. There were significant differences among sexes in the expression of GHR, and IGF mRNAs at the peripheral level in the liver and gonads. Females had lower hepatic GHRs and higher ovarian GHRs than male and sex-reversed fish. While the mRNA levels of IGF-1 were lower in the ovary, the levels of IGF-2 were higher compared with those in testes. A significant correlation between GHRs and IGFs was demonstrated in the liver and gonad (except for IGF-1). Multiple regression analysis showed a significant relationship between GH mRNA and both GHRs and IGFs in the liver and gonad. MT exerted androgenic and, to some extent, estrogenic effects on several physiological parameters and GH-IGF action.
Product Description
Supelco
17α-Methyltestosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
17α-Methyltestosterone, VETRANAL®, analytical standard
Sigma-Aldrich
17α-Methyltestosterone, ≥97.0% (HPLC)