Merck
  • Nitrogen incorporation in CH(4)-N(2) photochemical aerosol produced by far ultraviolet irradiation.
Nitrogen incorporation in CH(4)-N(2) photochemical aerosol produced by far ultraviolet irradiation.
Astrobiology (2012-04-24)
Melissa G Trainer, Jose L Jimenez, Yuk L Yung, Owen B Toon, Margaret A Tolbert
ABSTRACT
Nitrile incorporation into Titan aerosol accompanying hydrocarbon chemistry is thought to be driven by extreme UV wavelengths (λ<120 nm) or magnetospheric electrons in the outer reaches of the atmosphere. Far UV radiation (120-200 nm), which is transmitted down to the stratosphere of Titan, is expected to affect hydrocarbon chemistry only and not initiate the formation of nitrogenated species. We examined the chemical properties of photochemical aerosol produced at far UV wavelengths, using a high-resolution time-of-flight aerosol mass spectrometer (HR-ToF-AMS), which allows for elemental analysis of particle-phase products. Our results show that aerosol formed from CH(4)/N(2) photochemistry contains a surprising amount of nitrogen, up to 16% by mass, a result of photolysis in the far UV. The proportion of nitrogenated organics to hydrocarbon species is shown to be correlated with that of N(2) in the irradiated gas. The aerosol mass greatly decreases when N(2) is removed, which indicates that N(2) plays a major role in aerosol production. Because direct dissociation of N(2) is highly improbable given the immeasurably low cross section at the wavelengths studied, the chemical activation of N(2) must occur via another pathway. Any chemical activation of N(2) at wavelengths >120 nm is presently unaccounted for in atmospheric photochemical models. We suggest that reaction with CH radicals produced from CH(4) photolysis may provide a mechanism for incorporating N into the molecular structure of the aerosol. Further work is needed to understand the chemistry involved, as these processes may have significant implications for how we view prebiotic chemistry on early Earth and similar planets.
Sigma-Aldrich
Formamidine acetate salt, 99%
Sigma-Aldrich
Formamidine hydrochloride, 97%