Skip to Content
AT
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Amino acid spacing in isotachophoresi; on polyacrylamide gel: a critical evaluation.
Amino acid spacing in isotachophoresi; on polyacrylamide gel: a critical evaluation.
Analytical biochemistry (1979-04-01)
N Y Nguyen, A Chrambach
PMID
464281
Product Description
53319
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Pricing
89179
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Pricing
84959
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Pricing