Merck
  • Rapid detection of proteins in polyacrylamide electrophoresis gels with Direct Red 81 and Amido Black.
Rapid detection of proteins in polyacrylamide electrophoresis gels with Direct Red 81 and Amido Black.
Methods in molecular biology (Clifton, N.J.) (2012-05-16)
David Choveaux, Robert G E Krause, J P Dean Goldring
ABSTRACT
Proteins separated by SDS-polyacrylamide gel electrophoresis need to be stained with organic dyes to be visualized and to enable comparisons to be made between the intensity of protein bands to observe and determine differences in protein concentration. The standard protein staining is with Coomassie Blue R-250. Coomassie staining takes 1 h to complete. Direct Red 81 and Amido Black stain proteins within 10 min. This chapter describes Direct Red 81 and Amido Black staining in comparison to staining with Coomassie Blue R-250.
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade
Sigma-Aldrich
Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution
Sigma-Aldrich
Brilliant Blue R, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue R, pure
Sigma-Aldrich
Amido Black Staining Solution 2X, electrophoresis reagent
Sigma-Aldrich
Naphthol Blue Black, Dye content 80 %
Sigma-Aldrich
Direct Red 81, Dye content 50 %
Sigma-Aldrich
Naphthol Blue Black, Dye content ~50 %
Sigma-Aldrich
Naphthol Blue Black, BioReagent, suitable for electrophoresis