Merck
  • Cysteine racemization during the Fmoc solid phase peptide synthesis of the Nav1.7-selective peptide--protoxin II.
Cysteine racemization during the Fmoc solid phase peptide synthesis of the Nav1.7-selective peptide--protoxin II.
Journal of peptide science : an official publication of the European Peptide Society (2012-05-19)
Jae H Park, Kevin P Carlin, Gang Wu, Victor I Ilyin, Donald J Kyle
ABSTRACT
Protoxin II is biologically active peptide containing the inhibitory cystine knot motif. A synthetic version of the toxin was generated with standard Fmoc solid phase peptide synthesis. If N-methylmorpholine was used as a base during synthesis of the linear protoxin II, it was found that a significant amount of racemization (approximately 50%) was observed during the process of cysteine residue coupling. This racemization could be suppressed by substituting N-methylmorpholine with 2,4,6-collidine. The crude linear toxin was then air oxidized and purified. Electrophysiological assessment of the synthesized protoxin II confirmed its previously described interactions with voltage-gated sodium channels. Eight other naturally occurring inhibitory knot peptides were also synthesized using this same methodology. The inhibitory potencies of these synthesized toxins on Nav1.7 and Nav1.2 channels are summarized.
Sigma-Aldrich
4-Methylmorpholine, purum, ≥98.0% (GC)
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)