Merck
Sporotrichosis.
Clinics in dermatology (2012-06-12)
Elsa Vásquez-del-Mercado, Roberto Arenas, Carmen Padilla-Desgarenes
ABSTRACT
Sporotrichosis is the most common subcutaneous mycosis. It is usually acquired by traumatic inoculation, and it is caused by one of the species of the Sporothrix schenckii complex. More than 6 species, such as S schenckii sensu stricto, Sporothrix brasiliensis, Sporothrix globosa, Sporothrix mexicana, and Sporothrix albicans, have been identified by molecular techniques. The most common presentation is cutaneous disease, which is classified into fixed and lymphocutaneous forms. Osteoarticular, pulmonary, mucosal, disseminated, and systemic infections are less common and usually occur in immunosuppressed individuals. The diagnosis is suggested by biopsy specimen and confirmed by tissue culture. Itraconazole is considered the treatment of choice, although in some undeveloped countries potassium iodide is still used, owing to its safety and low cost. For systemic or disseminated cases, amphotericin B is the treatment of choice.
Sigma-Aldrich
Gold etchant, standard
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)