Merck
  • Efficient hydrodeoxygenation of biomass-derived ketones over bifunctional Pt-polyoxometalate catalyst.
Efficient hydrodeoxygenation of biomass-derived ketones over bifunctional Pt-polyoxometalate catalyst.
Chemical communications (Cambridge, England) (2012-06-12)
Mshari A Alotaibi, Elena F Kozhevnikova, Ivan V Kozhevnikov
ABSTRACT
Acidic heteropoly salt Cs(2.5)H(0.5)PW(12)O(40) doped with Pt nanoparticles is a highly active and selective catalyst for one-step hydrogenation of methyl isobutyl and diisobutyl ketones to the corresponding alkanes in the gas phase at 100 °C with 97-99% yield via metal-acid bifunctional catalysis.
Sigma-Aldrich
2,6-Dimethyl-4-heptanone, technical grade
Sigma-Aldrich
2,6-Dimethyl-4-heptanone, ≥99%
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Supelco
2-Hexanone, analytical standard
Supelco
4-Methyl-2-pentanone, analytical standard
Supelco
2-Hexanone, analytical standard
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%, FCC
Sigma-Aldrich
4-Methyl-2-pentanone, ACS reagent, ≥98.5%
Sigma-Aldrich
4-Methyl-2-pentanone, ≥99%
Sigma-Aldrich
2-Hexanone, reagent grade, 98%