Merck
  • Pd-catalyzed arylation of chlorotrifluoroethylene using arylboronic acids.
Pd-catalyzed arylation of chlorotrifluoroethylene using arylboronic acids.
Organic letters (2012-06-14)
Tetsuya Yamamoto, Tetsu Yamakawa
ABSTRACT
The palladium-catalyzed cross-coupling of chlorotrifluoroethylene and arylboronic acids proceeds in the presence of a base and H(2)O to provide α,β,β-trifluorostyrene derivatives in satisfactory yields.