Merck
  • Determination of 1-hydroxypyrene in human urine by HPLC with electrochemical detection at a boron-doped diamond film electrode.
Determination of 1-hydroxypyrene in human urine by HPLC with electrochemical detection at a boron-doped diamond film electrode.
Analytical and bioanalytical chemistry (2012-06-15)
Oksana Yosypchuk, Jiří Barek, Vlastimil Vyskočil
ABSTRACT
A high-performance liquid chromatographic method with electrochemical detection (HPLC-ED) at a boron-doped diamond film electrode with preliminary separation and preconcentration by solid-phase extraction (SPE) has been developed for the determination of 1-hydroxypyrene (1-HP) in human urine. 1-HP is among the most widely used biomarkers of exposure to polycyclic aromatic hydrocarbons. Optimal HPLC-ED conditions have been found: mobile phase methanol-0.05 mol L(-1) phosphate buffer pH 5.0 (80:20, v/v), detection potential +1,000 mV versus Ag/AgCl (3 mol L(-1) KCl), and flow rate 0.8 mL min(-1). For SPE, LiChrolut(®) RP-18 E cartridges were used. The extraction yield was (87.0 ± 5.8)% (n = 5). The concentration dependence of 1-HP was measured in the concentration range from 0.01 to 10 μmol L(-1) (2.18-2,180 μg L(-1)) using methanolic solutions resulting from the SPE pretreatment of spiked human urine samples. The limit of detection (signal-to-noise ratio 3) and the limit of quantification (signal-to-noise ratio 10) of the biomarker were 0.013 μmol L(-1) (2.84 μg L(-1)) and 0.043 μmol L(-1) (9.39 μg L(-1)), respectively, which is sufficient for its determination in the urine of persons exposed to polycyclic aromatic hydrocarbons.
Sigma-Aldrich
1-Aminopyrene, 97%
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
1-Hydroxypyrene, 98%
Sigma-Aldrich
1-Nitropyrene, ≥95%