Merck
  • Milnacipran inhibits glutamatergic N-methyl-D-aspartate receptor activity in spinal dorsal horn neurons.
Milnacipran inhibits glutamatergic N-methyl-D-aspartate receptor activity in spinal dorsal horn neurons.
Molecular pain (2012-06-22)
Tatsuro Kohno, Masafumi Kimura, Mika Sasaki, Hideaki Obata, Fumimasa Amaya, Shigeru Saito
ABSTRACT
Antidepressants, which are widely used for treatment of chronic pain, are thought to have antinociceptive effects by blockade of serotonin and noradrenaline reuptake. However, these drugs also interact with various receptors such as excitatory glutamatergic receptors. Thermal hyperalgesia was induced by intrathecal injection of NMDA in rats. Paw withdrawal latency was measured after intrathecal injection of antidepressants. The effects of antidepressants on the NMDA and AMPA-induced responses were examined in lamina II neurons of rat spinal cord slices using the whole-cell patch-clamp technique. The effects of milnacipran followed by application of NMDA on pERK activation were also investigated in the spinal cord. Intrathecal injection of milnacipran (0.1 μmol), but not citalopram (0.1 μmol) and desipramine (0.1 μmol), followed by intrathecal injection of NMDA (1 μg) suppressed thermal hyperalgesia. Milnacipran (100 μM) reduced the amplitude of NMDA (56 ± 3 %, 64 ± 5 % of control)-, but not AMPA (98 ± 5 %, 97 ± 5 % of control)-mediated currents induced by exogenous application and dorsal root stimulation, respectively. Citalopram (100 μM) and desipramine (30 μM) had no effect on the amplitude of exogenous NMDA-induced currents. The number of pERK-positive neurons in the group treated with milnacipran (100 μM), but not citalopram (100 μM) or desipramine (30 μM), followed by NMDA (100 μM) was significantly lower compared with the NMDA-alone group. The antinociceptive effect of milnacipran may be dependent on the drug's direct modulation of NMDA receptors in the superficial dorsal horn. Furthermore, in addition to inhibiting the reuptake of monoamines, glutamate NMDA receptors are also important for analgesia induced by milnacipran.
Product Description
Supelco
Desipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
WAY-100635 maleate salt, powder
Sigma-Aldrich
Desipramine hydrochloride, ≥98% (TLC), powder
Sigma-Aldrich
Citalopram hydrobromide, ≥98% (HPLC)
Desipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard